EMAIL THIS PAGE TO A FRIEND
International journal of nanomedicine
Tunable and noncytotoxic PET/SPECT-MRI multimodality imaging probes using colloidally stable ligand-free superparamagnetic iron oxide nanoparticles.
PMID 28184160
Abstract
Physiologically stable multimodality imaging probes for positron emission tomography/single-photon emission computed tomography (PET/SPECT)-magnetic resonance imaging (MRI) were synthesized using the superparamagnetic maghemite iron oxide (γ-Fe
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
427128
Gallium(III) chloride, beads, anhydrous, ≥99.999% trace metals basis
Cl3Ga