EMAIL THIS PAGE TO A FRIEND
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine
Phytochelatin 2 accumulates in roots of the seagrass Enhalus acoroides collected from sediment highly contaminated with lead.
PMID 28185077
Abstract
Phytochelatins (PCs), the heavy metal-binding peptides of plants, play a main function in heavy metal detoxification. In this study, Enhalus acoroides samples collected at six distinct seagrass beds from the coast of Khanh Hoa province, Viet Nam, were evaluated for their PCs. The contents of different PCs in each organ including leaf, rhizome, and root were determined by using HPLC analysis. Significant differences of PC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69480
N-Methyl-N-trimethylsilylacetamide, for GC derivatization, ≥97.0% (GC)
C6H15NOSi