EMAIL THIS PAGE TO A FRIEND
Chemico-biological interactions
Antitumor activity of paederosidic acid in human non-small cell lung cancer cells via inducing mitochondria-mediated apoptosis.
PMID 28185768
Abstract
This study was aimed to investigate antitumor activity of paederosidic acid (PA) in human non-small cell lung cancer cells and explore the related mechanisms. The anti-proliferative effects of PA on A549 cells were evaluated by MTT method and the IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU072971 MISSION® esiRNA, esiRNA human MAPK8 (esiRNA1)