EMAIL THIS PAGE TO A FRIEND
Journal of dermatological science
Intact wound repair activity of human mesenchymal stem cells after YM155 mediated selective ablation of undifferentiated human embryonic stem cells.
PMID 28185769
Abstract
Risk of teratoma formation during human pluripotent stem cell (hPSC)-based cell therapy is one of the technical hurdles that must be resolved before their wider clinical application. To this end, selective ablation of undifferentiated hPSCs has been achieved using small molecules whose application should be safe for differentiated cells derived from the hPSCs. However, the functional safety of such small molecules in the cells differentiated from hPSCs has not yet been extensively validated. We used the survivin inhibitor YM155, which induced highly selective cell death of hPSCs for ablating undifferentiated hESCs after differentiation to human mesenchymal stem cells (hMSCs) and examined whether hMSCs remained fully functional after being exposed by YM155. We demonstrated that human mesenchymal stem cells (hMSCs) derived from human embryonic stem cells (hESCs) remained fully functional in vitro and in vivo, while hESCs were selectively ablated. These results suggest that a single treatment with YM155 after differentiation of hMSCs would be a valid approach for teratoma-free cell therapy.
Related Materials