EMAIL THIS PAGE TO A FRIEND
Nitric oxide : biology and chemistry
S-Transnitrosation reactions of hydrogen sulfide (H
PMID 28185882
Abstract
Cysteine (CysSH) and its derivatives including N-acetylcysteine (NAC) and glutathione (GSH), and cysteine residues in proteins and enzymes are nitrosated with nitric oxide (NO) reaction products such as N
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
137030
Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate, 99%
C14H21NO4