EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Recruitment of PP1 to the centrosomal scaffold protein CEP192.
PMID 28188792
Abstract
Centrosomal protein of 192 kDa (CEP192) is a scaffolding protein that recruits the mitotic protein kinases Aurora A and PLK1 to the centrosome. Here we demonstrate that CEP192 also recruits the type one protein phosphatase (PP1) via a highly conserved KHVTF docking motif. The threonine of the KHVTF motif is phosphorylated during mitosis and protein kinase inhibition studies suggest this to be a PLK1-dependent process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33893
Microcystin-LR solution, 10 μg/mL in methanol, analytical standard
C49H74N10O12