EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
Water-soluble metalloporphyrinates with excellent photo-induced anticancer activity resulting from high tumor accumulation.
PMID 28189031
Abstract
To develop a water-soluble and tumor-targeted photosensitizer for photodynamic therapy (PDT), a porphyrin framework containing the metal ion gallium(III) was combined with platinum(II)-based groups to produce two new pentacationic metalloporphyrinates, Ga-4cisPtTPyP (5,10,15,20-tetrakis{cis-diammine-chloro-platinum(II)}(4-pyridyl)-porphyrinato gallium(III) hydroxide tetranitrate) and Ga-4transPtTPyP (5,10,15,20-tetrakis{trans-diammine-chloro-platinum(II)} (4-pyridyl)-porphyrinato gallium(III) hydroxide tetranitrate). Both complexes exhibited high singlet oxygen quantum yields (Φ