EMAIL THIS PAGE TO A FRIEND
Neurobiology of disease
Immunization with Bacillus Calmette-Guérin (BCG) alleviates neuroinflammation and cognitive deficits in APP/PS1 mice via the recruitment of inflammation-resolving monocytes to the brain.
PMID 28189498
Abstract
The immune system plays a crucial role in the progression of Alzheimer's disease (AD). Recently, immune-dependent cascade induced by systemic immune activation has been verified to play a beneficial role in AD mouse models. Here, we tested whether Bacillus Calmette-Guérin (BCG) immunization alters AD pathology and cognitive dysfunction in APP/PS1 AD mouse model, and with 4Aβ1-15 vaccination as positive control. It was found that BCG treatment reversed the cognitive decline to the extent observed in 4Aβ1-15 group, but did not reduce the β-amyloid (Aβ) burden in the brain. Then, we demonstrated the enhanced recruitment of inflammation-resolving monocytes across the choroid plexus and perivascular spaces to cerebral sites of plaque pathology in APP/PS1 mice immunized with BCG. Furthermore, elevated splenocyte Foxp3
Related Materials