EMAIL THIS PAGE TO A FRIEND
Experimental cell research
Lysophosphatidic acid signaling via LPA
PMID 28189636
Abstract
Lysophosphatidic acid (LPA) signaling via G protein-coupled LPA receptors exhibits a variety of biological effects, such as cell proliferation, motility and differentiation. The aim of this study was to evaluate the roles of LPA