EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Separation of betacyanins from purple flowers of Gomphrena globosa L. by ion-pair high-speed counter-current chromatography.
PMID 28190594
Abstract
Betacyanins, known as antioxidants and chemopreventive natural compounds with colourful properties, were extracted from purple flowers of Gomphrena globosa L. belonging to the Amaranthaceae family and separated for the first time by ion-pair high-speed counter-current chromatography (HSCCC). The pigments were detected by LC-DAD-ESI-MS/MS technique. Separation of betacyanins (300mg) by HSCCC was accomplished in four solvent systems: tert-butyl methyl ether - butanol - acetonitrile - water (0.7% and 1.0% HFBA - heptafluorobutyric acid - system I and III) and tert-butyl methyl ether - butanol - methanol - water (0.7% and 1.0% HFBA - system II and IV) (2:2:1:5, v/v/v/v) in the head-to-tail mode. The mobile phase (aqueous phase) was run at 2.0ml/min and the column rotation speed was 860rpm. The applied systems enabled to study the influence of HFBA concentration as well as systems polarity on betacyanins separation. Comparison of the systems containing 0.7% HFBA (systems I-II) demonstrates that the replacement of acetonitrile by methanol increases the resolution (R
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
443751
2,2,3,3,4,4,4-Heptafluorobutyl acrylate, 97%
C7H5F7O2