Experimental dermatology
Metformin alleviates ageing cellular phenotypes in Hutchinson-Gilford progeria syndrome dermal fibroblasts.
PMID 28192606
Metformin is a popular antidiabetic biguanide, which has been considered as a candidate drug for cancer treatment and ageing prevention. Hutchinson-Gilford progeria syndrome (HGPS) is a devastating disease characterized by premature ageing and severe age-associated complications leading to death. The effects of metformin on HGPS dermal fibroblasts remain largely undefined. In this study, we investigated whether metformin could exert a beneficial effect on nuclear abnormalities and delay senescence in fibroblasts derived from HGPS patients. Metformin treatment partially restored normal nuclear phenotypes, delayed senescence, activated the phosphorylation of AMP-activated protein kinase and decreased reactive oxygen species formation in HGPS dermal fibroblasts. Interestingly, metformin reduced the number of phosphorylated histone variant H2AX-positive DNA damage foci and suppressed progerin protein expression, compared to the control. Furthermore, metformin-supplemented aged mice showed higher splenocyte proliferation and mRNA expression of the antioxidant enzyme, superoxide dismutase 2 than the control mice. Collectively, our results show that metformin treatment alleviates the nuclear defects and premature ageing phenotypes in HGPS fibroblasts. Thus, metformin can be considered a promising therapeutic approach for life extension in HGPS.