EMAIL THIS PAGE TO A FRIEND
ACS applied materials & interfaces
Fluorene-Based Two-Dimensional Covalent Organic Framework with Thermoelectric Properties through Doping.
PMID 28192662
Abstract
Organic semiconductors have great potential as flexible thermoelectric materials. A fluorene-based covalent organic framework (FL-COF-1) was designed with the aim of creating an enhanced π-π interaction among the crystalline backbones. By the introduction of fluorene units into the frameworks, the FL-COF-1 had high thermal stability with a BET surface area over 1300 m
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D17106
2,7-Diaminofluorene, >97%
C13H12N2
49825
DMACA Reagent, for microbiology
C11H13NO