EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
4-hydroxybenzaldehyde-chitooligomers suppresses H
PMID 28192685
Abstract
Positive charges of chitooligomer (COS) enable COS to interact with negatively charged anionic groups on the cell surface resulting in an improvement in the biological activity of COS and its derivatives. In this study, 4-hydroxybenzaldehyde-COS (HB-COS) was synthesized and investigated for its abilities against H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
144088
4-Hydroxybenzaldehyde, 98%
C7H6O2