Analytical chemistry
Experimental Confirmation of Isotope Fractionation in Thiomolybdates Using Ion Chromatographic Separation and Detection by Multicollector ICPMS.
PMID 28192962
Molybdenum (98)Mo/(95)Mo isotope ratios are a sediment paleo proxy for the redox state of the ancient ocean. Under sulfidic conditions, no fractionation between seawater and sediment should be observed if molybdate (MoO4(2-)) is quantitatively transformed to tetrathiomolybdate (MoS4(2-)) and precipitated. However, quantum mechanical calculations previously suggested that incomplete sulfidation could be associated with substantial fractionation. To experimentally confirm isotope fractionation in thiomolybdates, a new approach for determination of isotope ratios of individual thiomolybdate species was developed that uses chromatography (HPLC-UV) to separate individual thiomolybdates, collecting each peak and analyzing isotope ratios with multicollector inductively coupled plasma mass spectrometry (MC-ICPMS). Using commercially available MoO4(2-) and MoS4(2-) standards, the method was evaluated and excellent reproducibility and accuracy were obtained. For species with longer retention times, complete chromatographic peaks had to be collected to avoid isotope fractionation within peaks. Isotope fractionation during formation of thiomolybdates could be experimentally proven for the first time in the reaction of MoO4(2-) with 20-fold or 50-fold excess of sulfide. The previously calculated isotope fractionation for MoS4(2-) was confirmed, and the result for MoO2S2(2-) was in the predicted range. Isotopic fractionation during MoS4(2-) transformation with pressurized air was dominated by kinetic fractionation. Further optimization and online-coupling of the HPLC-MC-ICPMS approach for determination of low concentrations in natural samples will greatly help to obtain more accurate species-selective isotope information.
Related Materials
Product #
Molecular Formula
Add to Cart
Ammonium tetrathiomolybdate, 99.97% trace metals basis