EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Evaluation of layered double hydroxide/graphene hybrid as a sorbent in membrane-protected stir-bar supported micro-solid-phase extraction for determination of organochlorine pesticides in urine samples.
PMID 28193470
Abstract
In this work, the potential of layered double hydroxide/graphene (LDH-G) hybrid as a sorbent for extraction and preconcentration of fifteen organochlorine pesticides (OCPs) in urine samples was evaluated. The LDH-G hybrid was synthesized by co-precipitation method and it was then characterized by Fourier transform infrared spectroscopy, scanning electron microscopy, transmission electron microscopy and X-ray diffraction. The sorbent was then employed in membrane-protected stir-bar supported micro-solid-phase extraction (SB-μ-SPE) of OCPs in urine samples. This extraction approach is highly suitable for the samples representing matrix complexity such as urine because the sorbent is effectively protected inside the membrane. The extracted samples were analyzed by gas chromatography mass spectrometry. The factors that affect the performance of SB-μ-SPE were suitably optimized. This method demonstrated good linearity with coefficients of determination up to 0.9996. The limits of detection ranged between 0.22 and 1.38ngmL
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
309400
Hydrazine solution, 35 wt. % in H2O
H4N2