EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
Mdr1a plays a crucial role in regulating the analgesic effect and toxicity of aconitine by altering its pharmacokinetic characteristics.
PMID 28193520
Abstract
Aconitine (AC) is the primary bioactive/toxic alkaloid in plants of the Aconitum species. Our previous study demonstrated that Mdr1 was involved in efflux of AC. However, the mechanism by which Mdr1 regulates the efficacy/toxicity of AC in vivo remains unclear. The present study aimed to determine the effects of Mdr1a on the efficacy/toxicity and pharmacokinetics of AC in wild-type and Mdr1a
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0634 Human TNNI3 / Troponin I, Cardiac Muscle ELISA Kit