Photodiagnosis and photodynamic therapy
In vitro evaluation of ruthenium complexes for photodynamic therapy.
PMID 28193566
Photodynamic therapy (PDT) is a promising anti-tumor treatment strategy. Photosensitizer is one of the most important components of PDT. In this work, the anticancer activities of PDT mediated by six new ruthenium porphyrin complexes were screened. The mechanisms of the most efficacious candidate were investigated. Photocytotoxicity of the six porphyrins was tested. The most promising complex, Rup-03, was further investigated using Geimsa staining, which indirectly detects reactive oxygen species (ROS) and subcellular localization. Mitochondrial membrane potential (MMP), cell apoptosis, DNA fragmentation, c-Myc gene expression, and telomerase activities were also assayed. Rup-03 and Rup-04 had the lowest IC50 values. Rup-03 had an IC50 value of 29.5±2.3μM in HepG2 cells and 59.0±6.1μM in RAW264.7 cells, while Rup-04 had an IC50 value of 40.0±3.8μM in SGC-7901 cells. The complexes also induced cellular morphological changes and impaired cellular ability to scavenge ROS, and accumulated preferentially in mitochondria and endoplasmic reticulum. Rup-03 reduced MMP levels, induced apoptosis, and repressed both c-Myc mRNA expression and telomerase activity in HepG2 cells. Among six candidates, Rup-03-mediated PDT is most effective against HepG2 and RAW264.7, with a similar efficacy as that of Rup-04-mediated PDT against SGC-7901 cells. Repression of ROS scavenging activities and c-Myc expression, which mediated DNA damage-induced cell apoptosis and repression of telomerase activity, respectively, were found to be involved in the anticancer mechanisms of Rup-03.