EMAIL THIS PAGE TO A FRIEND
Molecular psychiatry
Enhancing VTA Ca
PMID 28194001
Abstract
Genetic factors significantly influence susceptibility for substance abuse and mood disorders. Rodent studies have begun to elucidate a role of Ca