EMAIL THIS PAGE TO A FRIEND
Nutrition research and practice
Anti-inflammatory activity of a sulfated polysaccharide isolated from an enzymatic digest of brown seaweed
PMID 28194259
Abstract
CPs were precipitated from The polysaccharides from the Celluclast enzyme digest (CCP) showed the highest inhibition of NO production in LPS-stimulated RAW 264.7 cells (IC Our results suggest that CCP has anti-inflammatory activities and is a potential candidate for the formulation of a functional food ingredient or/and drug to treat inflammatory diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5631 Fucoidan from Fucus vesiculosus, Crude