EMAIL THIS PAGE TO A FRIEND
Scientific reports
Lipid droplet dynamics and insulin sensitivity upon a 5-day high-fat diet in Caucasians and South Asians.
PMID 28195217
Abstract
A 5-day High-Fat High-Calorie diet (HFHC-diet) reduces insulin-stimulated glucose disposal (Rd) in South Asian, but not Caucasian healthy lean males. We aimed to investigate if differences in myocellular lipid handling are underlying this differential response. A two-step hyperinsulinemic-euglycemic clamp and muscle biopsies were performed in 12 healthy lean Caucasian and South Asian males (BMI < 25 kg/m