EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Small Molecule Mixture Analysis by Heteronuclear NMR under Spin Diffusion Conditions in Viscous DMSO-Water Solvent.
PMID 28195380
Abstract
Spin diffusion in NMR occurs for small- and medium-sized molecules when their tumbling rate reduces in solution so that magnetization exchange by longitudinal cross relaxation becomes highly efficient. Composite DMSO-water viscous solvents were used for the first time to access the individual NMR spectra of a mixture components in spin diffusion conditions. The easy handling and high dissolution power of [D
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4003
Ala-Tyr
C12H16N2O4