EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Soft, Wet-Chemical Synthesis of Metastable Superparamagnetic Hexagonal Close-Packed Nickel Nanoparticles in Different Ionic Liquids.
PMID 28196305
Abstract
The microwave-induced decomposition of bis{N,N'-diisopropylacetamidinate}nickel(II) [Ni{MeC(NiPr)