EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Engineered holocytochrome
PMID 28196881
Abstract
Cytochrome
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12072
Benzamidine, ≥95.0%
C7H8N2