EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Determination of the positions of aluminum atoms introduced into SSZ-35 and the catalytic properties of the generated Brønsted acid sites.
PMID 28197567
Abstract
The positions of aluminum (Al) atoms in SSZ-35 together with the characteristics of the generated protons were investigated by