EMAIL THIS PAGE TO A FRIEND
Supportive care in cancer : official journal of the Multinational Association of Supportive Care in Cancer
Follow-up after curative treatment for colorectal cancer: longitudinal evaluation of patient initiated follow-up in the first 12 months.
PMID 28197848
Abstract
To compare patient-triggered follow-up (PTFU) for curatively treated colorectal cancer against traditional outpatient follow-up (OPFU). Questionnaires were mailed at four time points over one-year post-treatment to two prospectively-recruited cohorts: A, patients entering follow-up and receiving OPFU pre-implementation of PTFU; B, patients entering follow-up (FU) and receiving either OPFU (B1) or PTFU (B2) post-implementation of PTFU. Bi-variate tests were used to compare patient characteristics and outcomes eight months after entering follow-up (generic and cancer-specific quality of life (QoL), satisfaction). Regression analysis explored associations between follow-up model and outcomes. Resource implications and costs of models were compared. Patients in Cohort B1 were significantly more likely to have received chemotherapy (p < 0.001), radiotherapy (p < 0.05), and reported poorer QoL (p = 0.001). Having a longstanding co-morbid condition was the most important determinant of QoL (p < 0.001); model of care was not significant. Patients were satisfied with their follow-up care regardless of model. Health service costs were higher in PTFU over the first year CONCLUSIONS: PTFU is acceptable to patients with colorectal cancer and can be considered to be a realistic alternative to OPFU for clinically suitable patients. The initial costs are higher due to provision of a self-management (SM) programme and remote surveillance. Further research is needed to establish long-term outcomes and costs.