EMAIL THIS PAGE TO A FRIEND
Biotechnology and bioengineering
Parallel experimental design and multivariate analysis provides efficient screening of cell culture media supplements to improve biosimilar product quality.
PMID 28197999
Abstract
Rational and high-throughput optimization of mammalian cell culture media has a great potential to modulate recombinant protein product quality. We present a process design method based on parallel design-of-experiment (DoE) of CHO fed-batch cultures in 96-deepwell plates to modulate monoclonal antibody (mAb) glycosylation using medium supplements. To reduce the risk of losing valuable information in an intricate joint screening, 17 compounds were separated into five different groups, considering their mode of biological action. The concentration ranges of the medium supplements were defined according to information encountered in the literature and in-house experience. The screening experiments produced wide glycosylation pattern ranges. Multivariate analysis including principal component analysis and decision trees was used to select the best performing glycosylation modulators. Subsequent D-optimal quadratic design with four factors (three promising compounds and temperature shift) in shake tubes confirmed the outcome of the selection process and provided a solid basis for sequential process development at a larger scale. The glycosylation profile with respect to the specifications for biosimilarity was greatly improved in shake tube experiments: 75% of the conditions were equally close or closer to the specifications for biosimilarity than the best 25% in 96-deepwell plates. Biotechnol. Bioeng. 2017;114: 1448-1458. © 2017 Wiley Periodicals, Inc.