EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
A dioxidovanadium (V) complex of NNO-donor Schiff base as a selective inhibitor of protein tyrosine phosphatase 1B: Synthesis, characterization, and biological activities.
PMID 28199951
Abstract
A new dioxidovanadium (V) complex, VO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7653
p-Nitrophenyl Phosphate Liquid Substrate System, liquid
C6H4NNa2O6P