EMAIL THIS PAGE TO A FRIEND
Cancer research
Aberrant SYK Kinase Signaling Is Essential for Tumorigenesis Induced by TSC2 Inactivation.
PMID 28202529
Abstract
Somatic or germline mutations in the tuberous sclerosis complex (TSC) tumor suppressor genes are associated closely with the pathogenesis of lymphangioleiomyomatosis, a rare and progressive neoplastic disease that predominantly affects women in their childbearing years. Serum levels of the lymphangiogenic growth factor VEGF-D are elevated significantly in lymphangioleiomyomatosis. However, there are gaps in knowledge regarding VEGF-D dysregulation and its cellular origin in lymphangioleiomyomatosis. Here, we show that increased expression and activation of the tyrosine kinase Syk in TSC2-deficient cells and pulmonary nodules from lymphangioleiomyomatosis patients contributes to tumor growth. Syk kinase inhibitors blocked Syk signaling and exhibited potent antiproliferative activities in TSC2-deficient cells and an immunodeficient mouse xenograft model of lymphangioleiomyomatosis. In TSC2-deficient cells, Syk signaling increased the expression of monocyte chemoattractant protein MCP-1, which in peripheral blood mononuclear cells (PBMC) stimulated the production of VEGF-D. In clinical isolates of PBMCs from lymphangioleiomyomatosis patients, VEGF-D expression was elevated. Furthermore, levels of VEGF-D and MCP-1 in patient sera correlated positively with each other. Our results illuminate the basis for lymphangioleiomyomatosis growth and demonstrate the therapeutic potential of targeting Syk in this and other settings driven by TSC genetic mutation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU122051 MISSION® esiRNA, esiRNA human STAT3 (esiRNA1)