EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Exopolysaccharide from
PMID 28202619
Abstract
Commensal bacteria contribute to immune homeostasis in the gastrointestinal tract; however, the underlying mechanisms for this are not well understood. A single dose of exopolysaccharide (EPS) from the probiotic spore-forming bacterium