EMAIL THIS PAGE TO A FRIEND
Clinical endocrinology
Incomplete masculinization due to a deficiency of 17 beta-hydroxysteroid dehydrogenase: comparison of prepubertal and peripubertal siblings.
PMID 2820622
Abstract
Incomplete masculinization due to a deficiency of 17 beta-hydroxysteroid dehydrogenase (17 beta-HSD) was investigated in siblings aged 4 years (Case 1) and 12 years (Case 2). Diagnosis was based on increased ratios of androstenedione (A) to testosterone (T) in blood, and impaired reduction of A to T by 17 beta-HSD in vitro in the testes. Impairment was total in Case 2 but partial in Case 1. Case 2 also showed deficient conversion of dehydroepiandrosterone (DHA) to androstenediol and of oestrone to oestradiol by 17 beta-HSD which were normal in Case 1. Oxidation of T to A by 17 beta-HSD and conversion of 17 alpha-hydroxyprogesterone to A by 17,20 desmolase were normal in the testes of both siblings. 3 beta-HSD conversion of DHA to A was normal in Case 1, but markedly increased in Case 2. In contrast to testicular findings, 17 beta-HSD reduction of A to T in genital skin fibroblasts from Case 2 was normal and diagnosis would not have been possible from studies of measurements of this enzyme in skin. The severity of the testicular 17 beta-HSD deficiency in the peripubertal compared with the prepubertal sibling suggests either considerable intra-familial variation in the extent of the enzyme defect or that puberty may aggravate this disorder. The normal reductive action of 17 beta-HSD in skin, despite impaired action in testes, suggests involvement of more than one iso-enzyme.