EMAIL THIS PAGE TO A FRIEND
Drug metabolism and pharmacokinetics
Contribution of equilibrative nucleoside transporter (ENT) 2 to fluorouracil transport in rat placental trophoblast cells.
PMID 28209435
Abstract
Fluorouracil is used for treatment of breast cancer even in pregnant women, except during fetal organogenesis. The purpose of this study was to clarify the transport mechanism of fluorouracil at the rat placental barrier. Maternal-to-fetal transfer of [