EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Distance-dependent gradient in NMDAR-driven spine calcium signals along tapering dendrites.
PMID 28209776
Abstract
Neurons receive a multitude of synaptic inputs along their dendritic arbor, but how this highly heterogeneous population of synaptic compartments is spatially organized remains unclear. By measuring
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5500
L-(−)-Glucose, ≥99%
C6H12O6