EMAIL THIS PAGE TO A FRIEND
Talanta
Silica - Boronate affinity material for quick enrichment of intracellular nucleosides.
PMID 28213215
Abstract
Boronic acid modified materials have been widely used to adsorb nucleosides, but their adsorption capacities require further improvement. Most cis-diol containing biomolecules are in very low abundance along with interfering components in real samples, and need to be enriched specially. In this study, we synthesize a kind of silica absorbent modified with boronic acid derivative, using amorphous silica as raw material and obtaining high adsorption capacity for adenosine. In addition, the adsorption equilibrium can be completed within 10s and 1min for the desorption. Finally, the material was successfully applied to enrich nucleosides from cells and the spiked recoveries were found between 82.21% and 118.9%. The results showed that the prepared adsorbent has potential to effectively enrich cis-diol substances from cell samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
281778
(3-Aminopropyl)trimethoxysilane, 97%
C6H17NO3Si
I4125
Inosine, ≥99% (HPLC)
C10H12N4O5