EMAIL THIS PAGE TO A FRIEND
Talanta
Graphene quantum dots decorated with maleimide and zinc tetramaleimido-phthalocyanine: Application in the design of "OFF-ON" fluorescence sensors for biothiols.
PMID 28213216
Abstract
The fabrication of maleimide-derivatized graphene quantum dots (M-GQDs) and zinc phthalocyanine (2) as novel sensor probes for the selective detection of biothiols (cysteine, homocysteine or glutathione) through the rapid and specific Michael addition reaction between biothiols and the maleimide-derivatized probes is presented in this study. GQDs directly functionalized with maleimide units (M-GQDs) were synthesized and deployed for biothiols recognition following the principle of Michael addition. M-GQDs probe was found to be highly sensitive and selective towards biothiols detection in the nanomolar range in aqueous solution and at physiological pH (7.0). On the other hand, non-covalent interaction between pristine GQDs and novel zinc tetramaleimido-derivatized phthalocyanine resulted in the quenching of the pristine GQDs fluorescence emission which was switched back to the "ON" mode by Michael addition mechanism in the presence of biothiols. Tested relevant biomolecules did not interfere in the quantitative recognition of the biothiols. The probes showed to be highly sensitive, specific and selective for biothiols sensing in simulated real samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS011290
4-(2,5-dioxo-2,5-dihydro-pyrrol-1-yl)-benzoic acid, AldrichCPR
C11H7NO4