EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
Responses of soil methanogens, methanotrophs, and methane fluxes to land-use conversion and fertilization in a hilly red soil region of southern China.
PMID 28213705
Abstract
Changes in land-uses and fertilization are important factors regulating methane (CH