EMAIL THIS PAGE TO A FRIEND
Journal of cellular physiology
Chrysin induces death of prostate cancer cells by inducing ROS and ER stress.
PMID 28213961
Abstract
Chrysin is a natural flavone found in numerous plant extracts, honey, and propolis that has multiple biological activities including anti-cancer effects. Understanding of biological mechanisms mediated in response to chrysin in cancerous cells may provide novel insight into chemotherapeutic approaches with reduced side effects in cancers. In the present study, we investigated functional roles of chrysin in progression of prostate cancer cells using DU145 and PC-3 cell lines. The results showed that chrysin induced apoptosis of cells evidenced by DNA fragmentation and increasing the population of both DU145 and PC-3 cells in the sub-G