EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
Pharmacokinetic alterations of rhubarb anthraquinones in experimental colitis induced by dextran sulfate sodium in the rat.
PMID 28214059
Abstract
Rhubarb (Rhei Rhizoma et Radix) is used for the treatment of digestive diseases in traditional medicinal practice in China. Recent studies also support its beneficial activities in alleviating ulcerative colitis (UC). This study aimed to characterize the oral pharmacokinetics of rhubarb anthraquinones, the main bioactive components of this herb, in the experimental chronic colitis rat model induced by dextran sulfate sodium (DSS) and to identify the factors causing the pharmacokinetic alterations. Rats received drinking water (normal group) or 5% DSS for the first 7 days and 3% DSS for additional 14 days (UC group). On day 21 both groups received an oral dose of the rhubarb extract (equivalent to 5.0g crude drug/kg body weight). Plasma anthraquinone aglycones levels were determined directly by an LC-MS/MS method and the total of each anthraquinone (aglycone+conjugates) was quantified after β-glucuronidases hydrolysis. Rhubarb anthraquinones predominantly existed as conjugates in plasma samples from both groups and only free aloe-emodin, rhein and emodin were detected. Compared to the normal rats, both C The decreases of β-glucuronidases activity in DSS-induced chronic rat colitis should mainly account for the decreases in systemic exposure and abrogation of enterohepatic recirculation of most rhubarb anthraquinones after oral intake.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U5625
Uridine 5′-diphosphoglucuronic acid ammonium salt, 98-100%
C15H22N2O18P2 · xNH3