EMAIL THIS PAGE TO A FRIEND
Redox biology
Influence of oxygen partial pressure on the characteristics of human hepatocarcinoma cells.
PMID 28214706
Abstract
Most of the in vitro studies using liver cell lines have been performed under atmospheric oxygen partial pressure (21% O