EMAIL THIS PAGE TO A FRIEND
Pharmacognosy magazine
Obtaining a Dry Extract from the
PMID 28216886
Abstract
To obtain hydroalcoholic dried extract by nebulization and evaluate its antiulcerogenic potential. Plant material and hydroalcoholic extract were processed and analyzed for their physicochemical characteristics. A method using HPLC was validated to quantify coumarin and o-coumaric acid. Hydroalcoholic extract was spray dried and the powder obtained was characterized in terms of its physicochemical parameters and potential for antiulcerogenic activity. The analytical method proved to be selective, linear, precise, accurate, sensitive, and robust. This dried extract from Research and development of standardized dried extract of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H22809
2-Hydroxycinnamic acid, predominantly trans, 97%
C9H8O3