EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Integrative hepatoprotective efficacy comparison of raw and vinegar-baked Radix Bupleuri using nuclear magnetic resonance-based metabolomics.
PMID 28219798
Abstract
Radix Bupleuri (RB), with a Chinese name Chaihu, is one of the most popular Traditional Chinese herbal drug. It can be baked with vinegar to afford vinegar-baked Radix Bupleuri (VBRB), which is used in Traditional Chinese Medicine (TCM) for liver diseases treatment. In the present study, nuclear magnetic resonance-based metabolomic approach was used to compare the liver protective effect of RB and two types of VBRBs, which were prepared by two kinds of vinegar. The contents of 14 metabolites in the liver of carbon tetrachloride (CCl