EMAIL THIS PAGE TO A FRIEND
Dental survey
The value of a knowledgeable dental supply representative.
PMID 282200
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282200
Galangin, autophagy inducing flavonoid
C15H10O5