EMAIL THIS PAGE TO A FRIEND
Scientific reports
Parenteral administration of factor Xa/IIa inhibitors limits experimental aortic aneurysm and atherosclerosis.
PMID 28220880
Abstract
Intraluminal thrombus is a consistent feature of human abdominal aortic aneurysm (AAA). Coagulation factor Xa (FXa) catalyses FII to thrombin (FIIa). We examined the effect of FXa/FIIa inhibition on experimental aortic aneurysm in apolipoprotein E-deficient (ApoE