EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Bridgehead isomer effects in bis(phosphido)-bridged diiron hexacarbonyl proton reduction electrocatalysts.
PMID 28221379
Abstract
The influence of the substitution, orientation and structure of the phosphido bridges in [Fe
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
674389
Phenylphosphine, ca. 10% weight in hexanes
C6H7P