EMAIL THIS PAGE TO A FRIEND
Marine pollution bulletin
Tributyltin exposure increases mortality of nodavirus infected Japanese medaka Oryzias latipes larvae.
PMID 28222863
Abstract
We investigated the effect of combined exposure to nodavirus infection and TBT on medaka (Oryzias latipes). Medaka larvae were infected by immersion in medium containing nodavirus at titers of 10
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T50202
Tributyltin chloride, 96%
C12H27ClSn