EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Individual and combined effects of Aflatoxin B
PMID 28223122
Abstract
To understand the combinatorial toxicity of mycotoxins, we measured the effects of individual, binary and tertiary combinations of Aflatoxin B