Experimental cell research
Foxc1 and Foxc2 are necessary to maintain glomerular podocytes.
PMID 28223138
Foxc1 and Foxc2 (Foxc1/2) are transcription factors involved in many biological processes. In adult kidneys, expression of Foxc1/2 is confined to the glomerular epithelial cells, i.e., podocytes. To bypass embryonic lethality of Foxc1/2 null mice, mice ubiquitously expressing inducible-Cre (ROSA26-CreER(T2)) or mice expressing Cre in podocytes (Nephrin-Cre) were mated with floxed-Foxc1 and floxed-Foxc2 mice. The CreER(T2) was activated in adult mice by administrations of tamoxifen. Eight weeks after tamoxifen treatment, ROSA26-CreER(T2); Foxc1(+/flox); Foxc2(flox/flox) mice developed microalbuminuria, while ROSA26-Cre ER(T2); Foxc1(flox/flox); Foxc2(+/flox) mice had no microalbuminuria. The kidneys of conditional-Foxc1/2 null mice showed proteinaceous casts, protein reabsorption droplets in tubules and huge vacuoles in podocytes, indicating severe podocyte injury and massive proteinuria. Comparison of gene expression profiles revealed that Foxc1/2 maintain expression of genes necessary for podocyte function such as podocin and Cxcl12. In addition, mice with an innate podocyte-specific deletion of Foxc1/2 by Nephrin-Cre develop similar podocyte injury. These results demonstrate dose-dependence of Foxc1/2 gene in maintaining the podocyte with a more critical role for Foxc2 than Foxc1 and a critical role of Foxc1/2 in regulating expression of genes that maintain podocyte integrity.
Related Materials
Product #
Molecular Formula
Add to Cart
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
C26H28N3 · 0.5 C23H14O6 · xH2O