EMAIL THIS PAGE TO A FRIEND
Journal of dentistry
Poly(methyl methacrylate) with TiO
PMID 28223199
Abstract
The aim of this study was to obtain a Poly(methylmethacrylate) (PMMA)-TiO 0.2, 0.4, 0.6, 1, 2.5 by weight% of TiO SEM images and EDX results highlighted the presence of TiO Significant improvements in polymer characteristics and nice dispersion of the TiO Incorporation of TiO