EMAIL THIS PAGE TO A FRIEND
Scientific reports
Combination of ruthenium(II)-arene complex [Ru(η
PMID 28223694
Abstract
Ruthenium-based compounds show strong potential as anti-cancer drugs and are being investigated as alternatives to other well-established metal-based chemotherapeutics. The organometallic compound [Ru(η