EMAIL THIS PAGE TO A FRIEND
World journal of gastroenterology
Remote ischemic perconditioning prevents liver transplantation-induced ischemia/reperfusion injury in rats: Role of ROS/RNS and eNOS.
PMID 28223727
Abstract
To investigate the underlying mechanisms of the protective role of remote ischemic perconditioning (RIPerC) in rat liver transplantation. Sprague-Dawley rats were subjected to sham, orthotopic liver transplantation (OLT), ischemic postconditioning (IPostC) or RIPerC. After 3 h reperfusion, blood samples were taken for measurement of alanine aminotransferase, aspartate aminotransferase, creatinine (Cr) and creatinine kinase-myocardial band (CK-MB). The liver lobes were harvested for the following measurements: reactive oxygen species (ROS), H Compared with the OLT group, the grafts subjected to RIPerC showed significantly improved liver and remote organ functions ( Liver graft protection by RIPerC is similar to or better than that of IPostC, and involves inhibition of oxidative stress and up-regulation of the PI3K/Akt/eNOS/NO pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7389
3-Nitro-L-tyrosine, crystalline
C9H10N2O5