EMAIL THIS PAGE TO A FRIEND
Frontiers in cellular and infection microbiology
Heparin-binding Hemagglutinin of
PMID 28224118
Abstract
Airway epithelial cell is often the initial site of attack by pathogens, and cell death is commonly caused by internalization of